Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286371.1 | NP_001273300.1 | 6 | 703 | |
NM_001286372.1 | NP_001273301.1 | 5 | 608 | ||
NM_016485.4 | NP_057569.2 | 6 | 703 | ||
GenBank mRNA | AF060225.1 | - | 6 | 593 | |
AF141341.1 | - | 6 | 582 | ||
AF151062.1 | - | 6 | 600 | ||
AF271994.1 | - | 6 | 592 | ||
AJ420507.1 | - | 6 | 598 | ||
AK000051.1 | - | 6 | 592 | ||
AK130738.1 | - | 6 | 595 | ||
AK301376.1 | - | 5 | 511 | ||
AL136684.1 | - | 6 | 596 | ||
AY007093.1 | - | 5 | 407 | ||
BC005937.1 | - | 6 | 588 | 4081455 | |
BC006989.1 | - | 6 | 602 | 3961637 | |
BC013217.1 | - | 2 | 96 | ||
BC022536.1 | - | 6 | 602 | 4817057 | |
BM806661.1 | - | 6 | 503 | ||
CR457207.1 | - | 6 | 577 |