Gene Transcripts

Gene Symbol
TM6SF1
Entrez Gene ID
53346
Gene Name
transmembrane 6 superfamily member 1
Gene Aliases
Chromosome Location
Chr. 15: 83107572 - 83145403 on Build GRCh38
UniGene ID
Hs.513094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144903.1 NP_001138375.1 8 833
NM_023003.3 NP_075379.2 9 926
XM_011521679.2 XP_011519981.1 9 851
XM_011521680.2 XP_011519982.1 9 851
XM_017022320.1 XP_016877809.1 9 851
XM_017022321.1 XP_016877810.1 8 758
XM_017022322.1 XP_016877811.1 9 851
XM_017022323.1 XP_016877812.1 9 978
XM_017022324.1 XP_016877813.1 8 862
XM_017022325.1 XP_016877814.1 9 840
XM_017022326.1 XP_016877815.1 9 840
XM_017022327.1 XP_016877816.1 9 1032
XM_017022328.1 XP_016877817.1 6 601
XM_017022329.1 XP_016877818.1 6 591
XM_017022330.1 XP_016877819.1 6 591
XM_017022331.1 XP_016877820.1 6 591
GenBank mRNA AF255922.1 - 9 874
AK223246.1 - 9 871
AK292100.1 - 9 851
AK295568.1 - 9 815
BC032007.1 - 5 437
BC063487.1 - 6 606 5240735
DQ021909.1 - 8 724

View Full Product Details