Gene Transcripts

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
Chromosome Location
Chr. 11: 124884218 - 124897935 on Build GRCh38
UniGene ID
Hs.524121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301088.1 NP_001288017.1 5 646
NM_019055.5 NP_061928.4 5 852
XM_006718861.1 XP_006718924.1 5 785
GenBank mRNA AF361473.1 - 5 781
AK027852.1 - 5 751
AK074163.1 - 4 382
AK289769.1 - 5 751
AK302628.1 - 5 516
AY358083.1 - 5 703
BC014995.1 - 4 464
BC039602.1 - 5 740 5590503
BC111562.1 - 5 746 40068748
BC111748.1 - 5 746 40068752

View Full Product Details