Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301064.1 | NP_001287993.1 | 11 | 1041 | |
NM_018427.4 | NP_060897.3 | 12 | 1131 | ||
XM_005255376.1 | XP_005255433.1 | 10 | 906 | ||
XM_005255377.4 | XP_005255434.1 | 9 | 855 | ||
XM_011522540.2 | XP_011520842.1 | 7 | 543 | ||
XM_011522541.1 | XP_011520843.1 | 11 | 970 | ||
GenBank mRNA | AF227156.1 | - | 12 | 1097 | |
AJ272050.1 | - | 12 | 1051 | ||
AK225662.1 | - | 12 | 1107 | ||
AK295486.1 | - | 9 | 724 | ||
AK300253.1 | - | 11 | 950 | ||
AK302994.1 | - | 9 | 778 | ||
AK303405.1 | - | 8 | 624 | ||
AK304856.1 | - | 12 | 997 | ||
AK314769.1 | - | 12 | 1045 | ||
AK316520.1 | - | 11 | 955 | ||
BC036182.1 | - | 12 | 1046 | ||
BC071868.1 | - | 4 | 366 | ||
BC132688.1 | - | 12 | 1048 | 40146710 | |
BC132690.1 | - | 12 | 1048 | 40146712 | |
DA764092.1 | - | 4 | 334 |