Gene Transcripts
- Gene Symbol
- BIVM
- Entrez Gene ID
- 54841
- Gene Name
- basic, immunoglobulin-like variable motif containing
- Gene Aliases
- Chromosome Location
- Chr. 13: 102799049 - 102841538 on Build GRCh38
- UniGene ID
- Hs.288809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159596.1 |
NP_001153068.1
|
2, 3 |
597 |
|
NM_017693.3 |
NP_060163.2
|
4, 5 |
1281 |
|
GenBank mRNA |
AF411385.1 |
- |
4, 5 |
1281 |
|
AK000166.1 |
- |
2, 3 |
493 |
|
AK225229.1 |
- |
2, 3 |
493 |
|
AK290658.1 |
- |
4, 5 |
1150 |
|
AK304391.1 |
- |
3, 4 |
1035 |
|
BC020663.1 |
- |
2, 3 |
425 |
|
BC051813.2 |
- |
2, 3 |
406 |
|
BC075084.2 |
- |
2, 3 |
629 |
30915598 |
BC075085.2 |
- |
2, 3 |
629 |
30915386 |
BC112339.1 |
- |
2, 3 |
352 |
40053977 |
- Gene Symbol
- BIVM-ERCC5
- Entrez Gene ID
- 100533467
- Gene Name
- BIVM-ERCC5 readthrough
- Gene Aliases
- ERCC5-202
- Chromosome Location
- Chr. 13: 102807146 - 102876001 on Build GRCh38
- UniGene ID
- Hs.258429
- Species
- Homo sapiens
View Full Product Details