Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr. 17: 38800434 - 38825350 on Build GRCh38
UniGene ID
Hs.406223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017748.4 NP_060218.1 7, 8 1066
NR_073428.1 8, 9 1183
GenBank mRNA AK000298.1 - 7, 8 1016
AK225373.1 - 7, 8 1036
AK296115.1 - 8, 9 1148
AK316077.1 - 6, 7 1012
BC008833.2 - 7, 8 1052 4304656

View Full Product Details