Gene Transcripts

Gene Symbol
C1orf56
Entrez Gene ID
54964
Gene Name
chromosome 1 open reading frame 56
Gene Aliases
MENT
Chromosome Location
Chr. 1: 151047783 - 151051420 on Build GRCh38
UniGene ID
Hs.549171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017860.3 NP_060330.2 1 1023
GenBank mRNA AK000526.1 - 3 883
AK301712.1 - 1 1095
AK315680.1 - 1 1035
AY358436.1 - 1 1066
BC002469.2 - 1 1023 3347975

View Full Product Details