Gene Transcripts

Gene Symbol
BANK1
Entrez Gene ID
55024
Gene Name
B-cell scaffold protein with ankyrin repeats 1
Gene Aliases
BANK
Chromosome Location
Chr. 4: 101790482 - 102074812 on Build GRCh38
UniGene ID
Hs.480400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083907.2 NP_001077376.2 9 1388
NM_001127507.2 NP_001120979.2 8 1220
NM_017935.4 NP_060405.4 9 1619
XM_017008337.1 XP_016863826.1 9 1366
GenBank mRNA AB063170.1 - 8 1255
AK000713.1 - 9 1335
AK091523.1 - 13 1720
AK292133.1 - 9 1463
AL834291.1 - 9 1438
BC032241.1 - 9 1487 5214147
EU051376.1 - 8 958

View Full Product Details