Gene Transcripts
- Gene Symbol
- OXR1
- Entrez Gene ID
- 55074
- Gene Name
- oxidation resistance 1
- Gene Aliases
- Nbla00307, TLDC3
- Chromosome Location
- Chr. 8: 106270178 - 106752694 on Build GRCh38
- UniGene ID
- Hs.127286
- Species
- Homo sapiens
View Full Product Details