Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 53 | 9902 | |
XM_006721062.1 | XP_006721125.1 | 53 | 10040 | ||
XM_011545882.2 | XP_011544184.1 | 54 | 10480 | ||
XM_011545883.1 | XP_011544185.1 | 54 | 10248 | ||
XM_011545885.2 | XP_011544187.1 | 54 | 10480 | ||
XM_011545886.2 | XP_011544188.1 | 46 | 8855 | ||
XM_011545888.2 | XP_011544190.1 | 36 | 7299 | ||
XM_011545889.2 | XP_011544191.1 | 36 | 7263 | ||
XM_017023426.1 | XP_016878915.1 | 53 | 10345 | ||
XM_017023427.1 | XP_016878916.1 | 52 | 10237 | ||
XM_017023428.1 | XP_016878917.1 | 51 | 9901 | ||
XM_017023430.1 | XP_016878919.1 | 12 | 3922 | ||
XM_017023432.1 | XP_016878921.1 | 53 | 10040 | ||
GenBank mRNA | AF494040.1 | - | 53 | 9902 | |
AK125952.1 | - | 6 | 2255 | ||
AL096732.1 | - | 1 | 450 |