Gene Transcripts

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
Chromosome Location
Chr. 11: 126269040 - 126278132 on Build GRCh38
UniGene ID
Hs.317190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017547.3 NP_060017.1 2 317
NR_037648.1 2 336
XM_011542895.2 XP_011541197.1 2 414
XM_011542896.2 XP_011541198.1 2 203
XM_017018000.1 XP_016873489.1 2 292
XM_017018001.1 XP_016873490.1 2 416
XM_017018003.1 XP_016873492.1 2 292
XM_017018004.1 XP_016873493.1 2 414
XM_017018005.1 XP_016873494.1 1 1449
XM_017018006.1 XP_016873495.1 2 292
GenBank mRNA AF103801.1 - 2 215
AF447877.1 - 1 564
AK023987.1 - 2 186
AK295267.1 - 2 272
AK295428.1 - 2 186
AL136923.1 - 2 197
BC013902.2 - 2 173 3956972
DA056004.1 - 2 317

View Full Product Details