Gene Transcripts
- Gene Symbol
- FOXRED1
- Entrez Gene ID
- 55572
- Gene Name
- FAD dependent oxidoreductase domain containing 1
- Gene Aliases
- FP634, H17
- Chromosome Location
- Chr. 11: 126269040 - 126278132 on Build GRCh38
- UniGene ID
- Hs.317190
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017547.3 |
NP_060017.1
|
3, 4 |
575 |
|
NR_037647.1 |
|
2, 3 |
354 |
|
NR_037648.1 |
|
3, 4 |
594 |
|
XM_006718879.3 |
XP_006718942.1
|
2, 3 |
288 |
|
XM_006718881.3 |
XP_006718944.1
|
2, 3 |
351 |
|
XM_011542895.2 |
XP_011541197.1
|
3, 4 |
672 |
|
XM_017018000.1 |
XP_016873489.1
|
3, 4 |
550 |
|
XM_017018003.1 |
XP_016873492.1
|
3, 4 |
550 |
|
XM_017018004.1 |
XP_016873493.1
|
3, 4 |
672 |
|
XM_017018005.1 |
XP_016873494.1
|
2, 3 |
1905 |
|
XM_017018006.1 |
XP_016873495.1
|
3, 4 |
550 |
|
GenBank mRNA |
AF103801.1 |
- |
3, 4 |
473 |
|
AF447877.1 |
- |
2, 3 |
822 |
|
AK023987.1 |
- |
3, 4 |
444 |
|
AK295267.1 |
- |
3, 4 |
530 |
|
AK298010.1 |
- |
2, 3 |
223 |
|
AK298807.1 |
- |
3, 4 |
464 |
|
AK302039.1 |
- |
2, 3 |
223 |
|
AL136923.1 |
- |
3, 4 |
455 |
|
BC002910.2 |
- |
1, 2 |
52 |
|
BC013902.2 |
- |
3, 4 |
431 |
3956972 |
View Full Product Details