Gene Transcripts
- Gene Symbol
- DEPDC1
- Entrez Gene ID
- 55635
- Gene Name
- DEP domain containing 1
- Gene Aliases
- DEP.8, DEPDC1-V2, DEPDC1A, SDP35
- Chromosome Location
- Chr. 1: 68474152 - 68497221 on Build GRCh38
- UniGene ID
- Hs.445098
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114120.2 |
NP_001107592.1
|
9, 10 |
2158 |
|
NM_017779.5 |
NP_060249.2
|
8, 9 |
1306 |
|
GenBank mRNA |
AB281187.1 |
- |
9, 10 |
2014 |
|
AB281274.1 |
- |
8, 9 |
1162 |
|
AJ278112.1 |
- |
8, 9 |
1162 |
|
AK000361.1 |
- |
9, 10 |
2019 |
|
AK000490.1 |
- |
2, 3 |
483 |
|
AK022792.1 |
- |
1, 2 |
799 |
|
AK225735.1 |
- |
9, 10 |
2004 |
|
AK299324.1 |
- |
9, 10 |
2019 |
|
AL834208.1 |
- |
10, 11 |
1078 |
|
BC065304.1 |
- |
8, 9 |
1109 |
6156084 |
View Full Product Details