Gene Transcripts
- Gene Symbol
- DEPDC1
- Entrez Gene ID
- 55635
- Gene Name
- DEP domain containing 1
- Gene Aliases
- DEP.8, DEPDC1-V2, DEPDC1A, SDP35
- Chromosome Location
- Chr. 1: 68474152 - 68497221 on Build GRCh38
- UniGene ID
- Hs.445098
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114120.2 |
NP_001107592.1
|
6, 7 |
995 |
|
NM_017779.5 |
NP_060249.2
|
6, 7 |
995 |
|
GenBank mRNA |
AB281187.1 |
- |
6, 7 |
851 |
|
AB281274.1 |
- |
6, 7 |
851 |
|
AJ278112.1 |
- |
6, 7 |
851 |
|
AK000361.1 |
- |
6, 7 |
856 |
|
AK225735.1 |
- |
6, 7 |
841 |
|
AK299324.1 |
- |
6, 7 |
856 |
|
AL834208.1 |
- |
6, 7 |
477 |
|
BC003511.1 |
- |
6, 7 |
844 |
|
BC065304.1 |
- |
6, 7 |
798 |
6156084 |
View Full Product Details