Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr. 6: 485138 - 693141 on Build GRCh38
UniGene ID
Hs.484412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018303.5 NP_060773.3 19 2127
NR_073064.1 21 2289
XM_017011018.1 XP_016866507.1 20 2173
XM_017011019.1 XP_016866508.1 20 2335
XM_017011020.1 XP_016866509.1 20 2195
XM_017011021.1 XP_016866510.1 19 2431
XM_017011022.1 XP_016866511.1 19 2127
XM_017011023.1 XP_016866512.1 19 2127
XM_017011024.1 XP_016866513.1 20 2173
XM_017011025.1 XP_016866514.1 19 2127
XM_017011026.1 XP_016866515.1 19 2127
GenBank mRNA AJ414403.1 - 19 2051
AK001888.1 - 15 1542
AK056564.1 - 19 2095
AK302941.1 - 9 868
AK314628.1 - 19 2127
AL833209.1 - 17 1683
AL833213.1 - 19 2073
BC016918.1 - 16 1607
BC080542.1 - 21 2259 5479496

View Full Product Details