Gene Transcripts
- Gene Symbol
- KIZ
- Entrez Gene ID
- 55857
- Gene Name
- kizuna centrosomal protein
- Gene Aliases
- C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
- Chromosome Location
- Chr. 20: 21125980 - 21246628 on Build GRCh38
- UniGene ID
- Hs.187635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163022.1 |
NP_001156494.1
|
2, 3 |
295 |
|
NM_001276389.1 |
NP_001263318.1
|
1, 2 |
232 |
|
NM_018474.4 |
NP_060944.3
|
3, 4 |
458 |
|
XM_011529296.2 |
XP_011527598.1
|
3, 4 |
461 |
|
XM_011529297.2 |
XP_011527599.1
|
3, 4 |
450 |
|
XM_011529298.2 |
XP_011527600.1
|
3, 4 |
443 |
|
XM_011529299.2 |
XP_011527601.1
|
2, 3 |
290 |
|
XM_017027951.1 |
XP_016883440.1
|
4, 5 |
404 |
|
XM_017027952.1 |
XP_016883441.1
|
2, 3 |
290 |
|
XM_017027953.1 |
XP_016883442.1
|
4, 5 |
476 |
|
XM_017027954.1 |
XP_016883443.1
|
2, 3 |
290 |
|
GenBank mRNA |
AF451990.1 |
- |
3, 4 |
436 |
|
AK025476.1 |
- |
3, 4 |
389 |
|
AK293567.1 |
- |
1, 2 |
176 |
|
AK295355.1 |
- |
2, 3 |
194 |
|
BC039296.1 |
- |
3, 4 |
420 |
|
BC045826.1 |
- |
2, 3 |
182 |
5262165 |
BC065550.1 |
- |
1, 2 |
39 |
|
BC105093.1 |
- |
3, 4 |
361 |
8144096 |
BC113370.1 |
- |
3, 4 |
361 |
8322422 |
View Full Product Details