Gene Transcripts
- Gene Symbol
- STK31
- Entrez Gene ID
- 56164
- Gene Name
- serine/threonine kinase 31
- Gene Aliases
- SGK396, TDRD8
- Chromosome Location
- Chr. 7: 23710141 - 23832513 on Build GRCh38
- UniGene ID
- Hs.309767
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001260504.1 |
NP_001247433.1
|
23, 24 |
3212 |
|
NM_031414.4 |
NP_113602.2
|
23, 24 |
2936 |
|
NM_032944.3 |
NP_116562.2
|
23, 24 |
3180 |
|
NR_048542.1 |
|
23, 24 |
2928 |
|
XM_006715758.3 |
XP_006715821.1
|
21, 22 |
2316 |
|
XM_011515449.2 |
XP_011513751.2
|
23, 24 |
2862 |
|
XM_011515450.1 |
XP_011513752.1
|
23, 24 |
3183 |
|
XM_011515452.2 |
XP_011513754.2
|
21, 22 |
2329 |
|
GenBank mRNA |
AF285599.1 |
- |
23, 24 |
2832 |
|
AK131214.1 |
- |
14, 15 |
1638 |
|
AK292474.1 |
- |
24, 25 |
2828 |
|
BC059374.1 |
- |
23, 24 |
2843 |
5268051 |
View Full Product Details