Gene Transcripts
- Gene Symbol
- PRTFDC1
- Entrez Gene ID
- 56952
- Gene Name
- phosphoribosyl transferase domain containing 1
- Gene Aliases
- HHGP
- Chromosome Location
- Chr. 10: 24848607 - 24952644 on Build GRCh38
- UniGene ID
- Hs.405619
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282786.1 |
NP_001269715.1
|
2, 3 |
214 |
|
NM_020200.6 |
NP_064585.1
|
2, 3 |
214 |
|
GenBank mRNA |
AB209792.1 |
- |
2, 3 |
164 |
|
AF226056.1 |
- |
2, 3 |
162 |
|
AK021950.1 |
- |
2, 3 |
174 |
|
AK222674.1 |
- |
2, 3 |
173 |
|
AK294130.1 |
- |
2, 3 |
173 |
|
AK304055.1 |
- |
2, 3 |
173 |
|
AK309614.1 |
- |
2, 3 |
203 |
|
BC008662.2 |
- |
2, 3 |
176 |
3871911 |
BX471710.1 |
- |
2, 3 |
212 |
|
View Full Product Details