Gene Transcripts

Gene Symbol
PELI1
Entrez Gene ID
57162
Gene Name
pellino E3 ubiquitin protein ligase 1
Gene Aliases
Chromosome Location
Chr. 2: 64092652 - 64144471 on Build GRCh38
UniGene ID
Hs.7886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020651.3 NP_065702.2 6, 7 1160
XM_005264437.3 XP_005264494.1 5, 6 979
XM_011532994.2 XP_011531296.1 6, 7 1819
XM_011532996.2 XP_011531298.1 6, 7 2578
XM_011532997.2 XP_011531299.1 6, 7 1195
XM_017004520.1 XP_016860009.1 7, 8 1294
XM_017004521.1 XP_016860010.1 7, 8 1153
GenBank mRNA AF300987.1 - 5, 6 715
AF302505.1 - 5, 6 4738
AJ278859.1 - 5, 6 700
AK027668.1 - 3, 4 471
AK312441.1 - 6, 7 1109
AY364257.1 - 5, 6 700
BC050019.1 - 6, 7 1114 5296990
BC050533.1 - 6, 7 1096
BC063611.1 - 6, 7 1088 4992160
BX649152.1 - 2, 3 2016

View Full Product Details