Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300985.1 | NP_001287914.1 | 6 | 738 | |
NM_001300986.1 | NP_001287915.1 | 6 | 762 | ||
NM_001300987.1 | NP_001287916.1 | 7 | 894 | ||
NM_001300988.1 | NP_001287917.1 | 6 | 738 | ||
NM_001300989.1 | NP_001287918.1 | 6 | 738 | ||
NM_001300990.1 | NP_001287919.1 | 5 | 662 | ||
NM_021190.3 | NP_067013.1 | 6 | 738 | ||
NR_125356.1 | 6 | 630 | |||
NR_125357.1 | 6 | 738 | |||
XM_011541874.2 | XP_011540176.1 | 6 | 571 | ||
XM_011541875.2 | XP_011540177.1 | 6 | 584 | ||
XM_011541876.2 | XP_011540178.1 | 6 | 601 | ||
XM_017001951.1 | XP_016857440.1 | 5 | 601 | ||
GenBank mRNA | AB051232.1 | - | 6 | 560 | |
AB051233.1 | - | 6 | 504 | ||
AF176085.1 | - | 6 | 530 | ||
AF530580.1 | - | 6 | 533 | ||
AF530581.1 | - | 6 | 533 | ||
AK022716.1 | - | 1 | 44 | ||
AK092216.1 | - | 5 | 602 | ||
AK295384.1 | - | 6 | 551 | ||
AK299897.1 | - | 7 | 675 | ||
AK299922.1 | - | 6 | 576 | ||
BC016582.2 | - | 6 | 507 | 3872881 |