Gene Transcripts

Gene Symbol
RRAGD
Entrez Gene ID
58528
Gene Name
Ras related GTP binding D
Gene Aliases
RAGD, bA11D8.2.1
Chromosome Location
Chr. 6: 89363509 - 89412470 on Build GRCh38
UniGene ID
Hs.31712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021244.4 NP_067067.1 5 1150
XM_005248755.4 XP_005248812.1 5 1344
XM_011536022.2 XP_011534324.1 5 922
GenBank mRNA AF272036.1 - 5 933
AK289799.1 - 5 1075
AL137502.1 - 4 648
BC003088.1 - 5 812 3536088

View Full Product Details