Gene Transcripts

Gene Symbol
CELF6
Entrez Gene ID
60677
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
BRUNOL6
Chromosome Location
Chr. 15: 72284727 - 72320184 on Build GRCh38
UniGene ID
Hs.348342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172684.1 NP_001166155.1 10 1254
NM_001172685.1 NP_001166156.1 9 1027
NM_052840.4 NP_443072.3 11 1573
GenBank mRNA AF401233.1 - 11 1518
AF425606.1 - 11 1334
AK124858.1 - 10 1127
AK131098.1 - 13 2722
AK294382.1 - 9 1027
AK296004.1 - 10 1190
BC030835.1 - 11 1276 4793742
BC033838.2 - 11 1177 4214670

View Full Product Details