Gene Transcripts

Gene Symbol
XPNPEP3
Entrez Gene ID
63929
Gene Name
X-prolyl aminopeptidase 3
Gene Aliases
APP3, ICP55, NPHPL1
Chromosome Location
Chr. 22: 40857081 - 40932819 on Build GRCh38
UniGene ID
Hs.529163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022098.3 NP_071381.1 9, 10 1464
GenBank mRNA AK313770.1 - 9, 10 1425
AL365514.1 - 9, 10 1447
AL834310.1 - 10, 11 1506
BC001681.2 - 9, 10 1431 2823656
BC004989.1 - 9, 10 1431 2823656
BX648018.1 - 9, 10 1459
CR456442.1 - 9, 10 1400
CR457396.1 - 9, 10 1363

View Full Product Details