Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031713.3 | NP_001026883.1 | 2 | 649 | |
XM_011514802.1 | XP_011513104.1 | 2 | 652 | ||
GenBank mRNA | AK024611.1 | - | 2 | 658 | |
BC008234.1 | - | 2 | 404 | ||
BC016850.1 | - | 2 | 623 | 3886144 |