Gene Transcripts

Gene Symbol
NOD2
Entrez Gene ID
64127
Gene Name
nucleotide binding oligomerization domain containing 2
Gene Aliases
ACUG, BLAU, CARD15, CD, CLR16.3, IBD1, NLRC2, NOD2B, PSORAS1
Chromosome Location
Chr. 16: 50693581 - 50733077 on Build GRCh38
UniGene ID
Hs.592072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293557.1 NP_001280486.1 4 2573
NM_022162.2 NP_071445.1 5 2613
XM_005256084.3 XP_005256141.1 5 2517
XM_006721242.3 XP_006721305.1 5 2517
XM_011523259.2 XP_011521561.1 3 2329
XM_017023535.1 XP_016879024.1 3 3887
XM_017023536.1 XP_016879025.1 4 2044
XM_017023537.1 XP_016879026.1 3 1938
XM_017023538.1 XP_016879027.1 4 2039
GenBank mRNA AF178930.1 - 5 2613
AY187233.1 - 2 226
AY187234.1 - 2 254
AY187239.1 - 2 226
AY187241.1 - 2 226

View Full Product Details