Gene Transcripts

Gene Symbol
SMAP2
Entrez Gene ID
64744
Gene Name
small ArfGAP2
Gene Aliases
SMAP1L
Chromosome Location
Chr. 1: 40373706 - 40423326 on Build GRCh38
UniGene ID
Hs.15200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198978.1 NP_001185907.1 10 1156
NM_001198979.1 NP_001185908.1 10 1240
NM_001198980.1 NP_001185909.1 10 1171
NM_022733.2 NP_073570.1 10 1598
XM_011541960.1 XP_011540262.1 11 1605
XM_017002093.1 XP_016857582.1 7 1297
GenBank mRNA AK294675.1 - 5 880
AK298702.1 - 10 1136
AK303975.1 - 10 1144
AK308280.1 - 10 1241
AK313105.1 - 10 1545
AK316218.1 - 10 1171
AL133206.1 - 10 1112
AL137764.1 - 10 1125
BC014512.1 - 8 937
BC021133.1 - 10 1248 4855320

View Full Product Details