Gene Transcripts

Gene Symbol
MARCH7
Entrez Gene ID
64844
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
AXO, AXOT, MARCH-VII, RNF177
Chromosome Location
Chr. 2: 159712457 - 159768583 on Build GRCh38
UniGene ID
Hs.529272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282805.1 NP_001269734.1 7 1265
NM_001282806.1 NP_001269735.1 4 969
NM_001282807.1 NP_001269736.1 4 1012
NM_022826.3 NP_073737.1 5 1137
XM_005246773.2 XP_005246830.1 5 1160
XM_005246774.1 XP_005246831.1 5 1160
XM_017004718.1 XP_016860207.1 5 1137
XM_017004719.1 XP_016860208.1 4 995
XM_017004720.1 XP_016860209.1 4 957
XM_017004721.1 XP_016860210.1 4 987
GenBank mRNA AK022973.1 - 5 1095
AK222880.1 - 5 1087
AK292836.1 - 7 1233
AK299051.1 - 4 937
AK302347.1 - 4 961
AK310238.1 - 4 968
AK316382.1 - 4 969
AL833651.1 - 1 138
BC065014.1 - 5 1061 6169288

View Full Product Details