Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023080.2 | NP_075568.1 | 4, 5 | 598 | |
GenBank mRNA | AK024642.1 | - | 4, 5 | 596 | |
BC010001.2 | - | 4, 5 | 558 | 4135341 | |
BC015181.1 | - | 3, 4 | 359 | ||
BX648996.1 | - | 3, 4 | 658 | ||
CR457330.1 | - | 3, 4 | 544 |