Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023080.2 | NP_075568.1 | 5 | 681 | |
GenBank mRNA | AK024642.1 | - | 5 | 679 | |
BC010001.2 | - | 5 | 641 | 4135341 | |
BC015181.1 | - | 4 | 442 | ||
BX648996.1 | - | 4 | 741 | ||
CR457330.1 | - | 4 | 627 |