Gene Transcripts

Gene Symbol
NUP37
Entrez Gene ID
79023
Gene Name
nucleoporin 37
Gene Aliases
p37
Chromosome Location
Chr. 12: 102074189 - 102120139 on Build GRCh38
UniGene ID
Hs.444276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024057.3 NP_076962.2 3 371
GenBank mRNA AF514994.1 - 2 225
AK026271.1 - 2 206
AK296733.1 - 3 329
AK310619.1 - 3 331
BC000861.2 - 2 289 3459639
DC420994.1 - 3 372

View Full Product Details