Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024592.4 | NP_078868.1 | 4 | 717 | |
XM_017008601.1 | XP_016864090.1 | 4 | 492 | ||
GenBank mRNA | AK023414.1 | - | 4 | 697 | |
BC002480.2 | - | 4 | 693 | 3350037 | |
CR457312.1 | - | 4 | 601 |