Gene Transcripts

Gene Symbol
CLIP4
Entrez Gene ID
79745
Gene Name
CAP-Gly domain containing linker protein family member 4
Gene Aliases
RSNL2
Chromosome Location
Chr. 2: 29097676 - 29183813 on Build GRCh38
UniGene ID
Hs.122927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287527.1 NP_001274456.1 2 263
NM_001287528.1 NP_001274457.1 2 299
NM_024692.5 NP_078968.3 2 267
NR_109844.1 2 299
XM_005264562.2 XP_005264619.1 2 294
XM_005264563.1 XP_005264620.1 2 299
XM_006712104.2 XP_006712167.1 2 339
XM_006712105.3 XP_006712168.1 2 181
XM_011533109.1 XP_011531411.1 2 47
XM_017004958.1 XP_016860447.1 2 126
XM_017004959.1 XP_016860448.1 2 181
GenBank mRNA AB209042.1 - 2 1878
AF433661.1 - 2 275
AK057267.1 - 2 300
AK291595.1 - 2 344
AK302325.1 - 2 249
AL834443.1 - 2 241
BC013862.1 - 2 247
BC126167.1 - 1 37 8991883
BC136331.1 - 1 37 9020318
BI600762.1 - 2 272
DC317668.1 - 2 299

View Full Product Details