Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102396.1 | NP_001095866.1 | 2 | 323 | |
NM_025073.2 | NP_079349.2 | 2 | 311 | ||
NR_049741.1 | 2 | 373 | |||
GenBank mRNA | AK024821.1 | - | 2 | 301 | |
AK126316.1 | - | 2 | 321 | ||
AK225734.1 | - | 2 | 302 | ||
BC005934.1 | - | 2 | 281 | 4249034 | |
CR749219.1 | - | 2 | 314 | ||
CR936771.1 | - | 2 | 314 | ||
DB459255.1 | - | 2 | 141 |