Gene Transcripts

Gene Symbol
TMEM254
Entrez Gene ID
80195
Gene Name
transmembrane protein 254
Gene Aliases
C10orf57, bA369J21.6
Chromosome Location
Chr. 10: 80078646 - 80092552 on Build GRCh38
UniGene ID
Hs.169982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270367.1 NP_001257296.1 4 404
NM_001270368.1 NP_001257297.1 4 417
NM_001270369.1 NP_001257298.1 5 456
NM_001270370.1 NP_001257299.1 6 463
NM_001270371.1 NP_001257300.1 5 478
NM_001270372.1 NP_001257301.1 4 396
NM_001270373.1 NP_001257302.1 5 435
NM_001270374.1 NP_001257303.1 5 785
NM_025125.3 NP_079401.2 4 383
NR_072984.1 5 400
NR_072985.1 5 407
NR_072986.1 4 305
NR_072987.1 5 357
XM_006717978.2 XP_006718041.1 5 735
XM_017016679.1 XP_016872168.1 6 575
GenBank mRNA AK023325.1 - 4 359
AK222536.1 - 4 364
AK300490.1 - 4 414
AK309425.1 - 5 682
AK309980.1 - 4 392
AK310160.1 - 5 365
AK311563.1 - 5 373
AL832397.1 - 4 280
AL832483.1 - 4 391
BC022252.1 - 4 359 4668550
BP204746.1 - 6 432
BP293793.1 - 5 410
BX537803.1 - 5 365
CR457310.1 - 4 329
CX756014.1 - 6 419
DA263618.1 - 5 315
DA573956.1 - 5 416
DA930892.1 - 4 372
DB485367.1 - 5 407

View Full Product Details