Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077484.1 | NP_001070952.1 | 3 | 798 | |
NM_001278387.1 | NP_001265316.1 | 4 | 1260 | ||
NM_001278388.1 | NP_001265317.1 | 2 | 1022 | ||
NM_001278389.1 | NP_001265318.1 | 3 | 287 | ||
NM_001278390.1 | NP_001265319.1 | 3 | 1000 | ||
NM_030674.3 | NP_109599.3 | 3 | 1137 | ||
XM_011538784.2 | XP_011537086.1 | 3 | 522 | ||
XM_011538786.2 | XP_011537088.1 | 3 | 522 | ||
XM_011538787.2 | XP_011537089.1 | 3 | 522 | ||
XM_017019990.1 | XP_016875479.1 | 3 | 752 | ||
GenBank mRNA | AF247166.1 | - | 3 | 483 | |
AF271070.1 | - | 3 | 381 | ||
AK027825.1 | - | 3 | 287 | ||
AK074949.1 | - | 1 | 45 | ||
AK225639.1 | - | 3 | 375 | ||
AK225778.1 | - | 3 | 397 | ||
AK309410.1 | - | 3 | 714 | ||
AK313731.1 | - | 1 | 88 | ||
BC010620.1 | - | 3 | 344 | 3871101 | |
BX647547.1 | - | 4 | 832 | ||
BX648495.1 | - | 3 | 725 | ||
BX648676.1 | - | 2 | 592 |