Gene Transcripts

Gene Symbol
CRISPLD1
Entrez Gene ID
83690
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Gene Aliases
CRISP-10, CRISP10, LCRISP1
Chromosome Location
Chr. 8: 74984473 - 75034558 on Build GRCh38
UniGene ID
Hs.436542
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286777.1 NP_001273706.1 8 931
NM_001286778.1 NP_001273707.1 9 1047
NM_031461.5 NP_113649.1 10 1559
GenBank mRNA AF142573.1 - 10 1155
AF329197.1 - 9 1134
AK304034.1 - 9 1035
AK304350.1 - 8 912
AK314048.1 - 10 1315
AL834301.1 - 10 1291
AY358564.1 - 10 1517
BC020514.1 - 10 1424 3856615

View Full Product Details