Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322831.1 | NP_001309760.1 | 13, 14 | 1588 | |
NM_001322834.1 | NP_001309763.1 | 13, 14 | 1588 | ||
NM_031913.4 | NP_114119.2 | 13, 14 | 1588 | ||
NR_136409.1 | 13, 14 | 1588 | |||
XM_017007307.1 | XP_016862796.1 | 12, 13 | 1040 | ||
GenBank mRNA | AK125080.1 | - | 13, 14 | 1546 | |
AK289530.1 | - | 13, 14 | 1545 | ||
BC037292.1 | - | 13, 14 | 1580 | 4837811 | |
DQ993202.1 | - | 13, 14 | 1362 |