Gene Transcripts

Gene Symbol
RSPH3
Entrez Gene ID
83861
Gene Name
radial spoke 3 homolog
Gene Aliases
CILD32, RSHL2, RSP3, dJ111C20.1
Chromosome Location
Chr. 6: 158947012 - 159000432 on Build GRCh38
UniGene ID
Hs.154628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031924.4 NP_114130.3 4, 5 1099
XM_017011347.1 XP_016866836.1 3, 4 420
GenBank mRNA AF353618.1 - 4, 5 698
AK057931.1 - 4, 5 1120
BC050604.1 - 4, 5 1099 6161669

View Full Product Details