Gene Transcripts

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
Chromosome Location
Chr. 11: 13387996 - 13463297 on Build GRCh38
UniGene ID
Hs.332382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297741.1 NP_001284670.1 4, 5 790
NM_001297742.1 NP_001284671.1 4, 5 980
NM_032320.6 NP_115696.2 5, 6 948
XM_017018405.1 XP_016873894.1 4, 5 1959
XM_017018406.1 XP_016873895.1 5, 6 938
XM_017018407.1 XP_016873896.1 5, 6 918
XM_017018408.1 XP_016873897.1 5, 6 918
XM_017018409.1 XP_016873898.1 4, 5 785
XM_017018410.1 XP_016873899.1 4, 5 777
XM_017018411.1 XP_016873900.1 4, 5 777
GenBank mRNA AF329102.2 - 5, 6 948
AK292392.1 - 5, 6 873
AK294259.1 - 4, 5 769
AK294768.1 - 4, 5 695
AK298226.1 - 4, 5 665
AK310935.1 - 4, 5 673
AK316163.1 - 4, 5 980
AL832981.1 - 5, 6 929
AY221959.2 - 5, 6 991
BC005071.1 - 5, 6 910 3531347
BC062633.1 - 1, 2 58

View Full Product Details