Gene Transcripts
- Gene Symbol
- FRMD5
- Entrez Gene ID
- 84978
- Gene Name
- FERM domain containing 5
- Gene Aliases
- Chromosome Location
- Chr. 15: 43870761 - 44195294 on Build GRCh38
- UniGene ID
- Hs.578544
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286490.1 |
NP_001273419.1
|
14, 15 |
1310 |
|
NM_001286491.1 |
NP_001273420.1
|
14, 15 |
1694 |
|
NM_001322949.1 |
NP_001309878.1
|
13, 14 |
1375 |
|
NM_001322950.1 |
NP_001309879.1
|
13, 14 |
1375 |
|
NM_001322951.1 |
NP_001309880.1
|
12, 13 |
1270 |
|
NM_032892.4 |
NP_116281.2
|
13, 14 |
1375 |
|
NR_104455.1 |
|
14, 15 |
1691 |
|
XM_005254730.3 |
XP_005254787.1
|
13, 14 |
1280 |
|
GenBank mRNA |
AK090572.1 |
- |
13, 14 |
1307 |
|
AK123018.1 |
- |
14, 15 |
1310 |
|
AK290013.1 |
- |
2, 3 |
169 |
|
BC043486.2 |
- |
14, 15 |
1441 |
|
BC053647.1 |
- |
13, 14 |
1312 |
5578205 |
View Full Product Details