Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133367.4 | NP_588608.1 | 2 | 940 | |
GenBank mRNA | AF313619.1 | - | 1 | 372 | |
AF347029.1 | - | 2 | 930 | ||
AK093603.1 | - | 1 | 556 | ||
AK128085.1 | - | 1 | 1160 | ||
AK315088.1 | - | 2 | 918 | ||
AY424286.1 | - | 3 | 943 | ||
BC030664.2 | - | 2 | 915 | 5260230 |