Gene Transcripts
- Gene Symbol
- DOCK7
- Entrez Gene ID
- 85440
- Gene Name
- dedicator of cytokinesis 7
- Gene Aliases
- EIEE23, ZIR2
- Chromosome Location
- Chr. 1: 62454726 - 62688368 on Build GRCh38
- UniGene ID
- Hs.744927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271999.1 |
NP_001258928.1
|
5, 6 |
621 |
|
NM_001272000.1 |
NP_001258929.1
|
5, 6 |
621 |
|
NM_001272001.1 |
NP_001258930.1
|
5, 6 |
621 |
|
NM_001272002.1 |
NP_001258931.1
|
5, 6 |
621 |
|
NM_033407.3 |
NP_212132.2
|
5, 6 |
621 |
|
XM_005271292.2 |
XP_005271349.1
|
5, 6 |
611 |
|
XM_011542326.2 |
XP_011540628.1
|
5, 6 |
611 |
|
XM_011542327.2 |
XP_011540629.1
|
5, 6 |
611 |
|
XM_011542328.2 |
XP_011540630.1
|
5, 6 |
611 |
|
XM_011542330.2 |
XP_011540632.1
|
5, 6 |
611 |
|
XM_017002639.1 |
XP_016858128.1
|
5, 6 |
611 |
|
XM_017002640.1 |
XP_016858129.1
|
5, 6 |
611 |
|
GenBank mRNA |
AK055401.1 |
- |
5, 6 |
551 |
|
AK292640.1 |
- |
5, 6 |
611 |
|
DQ118679.1 |
- |
2, 3 |
193 |
|
DQ118680.1 |
- |
2, 3 |
193 |
|
DQ309763.1 |
- |
2, 3 |
193 |
|
DQ341187.1 |
- |
5, 6 |
528 |
|
View Full Product Details