Gene Transcripts

Gene Symbol
CNTNAP4
Entrez Gene ID
85445
Gene Name
contactin associated protein like 4
Gene Aliases
CASPR4
Chromosome Location
Chr. 16: 76277217 - 76563610 on Build GRCh38
UniGene ID
Hs.461389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322178.1 NP_001309107.1 10, 11 2048
NM_001322179.1 NP_001309108.1 11, 12 2198
NM_001322180.1 NP_001309109.1 10, 11 2004
NM_001322181.1 NP_001309110.1 11, 12 2195
NM_001322187.1 NP_001309116.1 12, 13 2263
NM_001322188.1 NP_001309117.1 11, 12 2198
NM_001322189.1 NP_001309118.1 12, 13 2457
NM_001322190.1 NP_001309119.1 10, 11 2054
NM_001322191.1 NP_001309120.1 10, 11 1999
NM_033401.4 NP_207837.2 11, 12 2198
NM_138994.4 NP_620481.2 10, 11 1592
NR_136210.1 11, 12 2198
NR_136213.1 11, 12 2193
NR_136216.1 13, 14 2381
XM_011523403.2 XP_011521705.1 11, 12 1736
XM_017023805.1 XP_016879294.1 11, 12 1927
GenBank mRNA AB051550.2 - 11, 12 2137
AB100092.1 - 10, 11 1608
AB100093.1 - 9, 10 1533
AB100094.1 - 11, 12 1752
AF463518.1 - 10, 11 1900
AK057327.1 - 11, 12 1722

View Full Product Details