Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
Chromosome Location
Chr. 21: 42062959 - 42142996 on Build GRCh38
UniGene ID
Hs.242520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004416.2 NP_001004416.2 20, 21 3699
NM_001199527.1 NP_001186456.1 19, 20 4263
NM_001199528.2 NP_001186457.2 20, 21 3879
NM_173568.3 NP_775839.3 19, 20 4083
XM_011529797.1 XP_011528099.1 17, 18 3691
XM_017028506.1 XP_016883995.1 19, 20 4078
XM_017028507.1 XP_016883996.1 19, 20 7007
XM_017028508.1 XP_016883997.1 9, 10 1862
GenBank mRNA AB051810.1 - 19, 20 4263
AB051811.1 - 20, 21 3879
AK093654.1 - 5, 6 941
AK124223.1 - 6, 7 1443
AY771619.1 - 19, 20 4083
AY771620.1 - 20, 21 3699

View Full Product Details