Gene Transcripts

Gene Symbol
CREB3L1
Entrez Gene ID
90993
Gene Name
cAMP responsive element binding protein 3 like 1
Gene Aliases
OASIS
Chromosome Location
Chr. 11: 46277638 - 46321422 on Build GRCh38
UniGene ID
Hs.405961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052854.3 NP_443086.1 12 2019
GenBank mRNA AB063321.1 - 12 1640
AF055009.1 - 4 1139
AK075538.1 - 7 790
AK314072.1 - 12 1997
BC014097.1 - 12 1662 4546950
BC015781.1 - 12 1648 4856946

View Full Product Details