Gene Transcripts

Gene Symbol
ACY3
Entrez Gene ID
91703
Gene Name
aminoacylase 3
Gene Aliases
ACY-3, HCBP1
Chromosome Location
Chr. 11: 67642551 - 67650829 on Build GRCh38
UniGene ID
Hs.126265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080658.1 NP_542389.1 4 433
XM_017018549.1 XP_016874038.1 5 624
XM_017018550.1 XP_016874039.1 5 658
XM_017018551.1 XP_016874040.1 5 579
GenBank mRNA AF359506.1 - 3 352
AY032593.1 - 413
AY040761.1 - 4 413
AY169233.1 - 4 433
BC008689.1 - 4 433 3853707

View Full Product Details