Gene Transcripts
- Gene Symbol
- HNRNPLL
- Entrez Gene ID
- 92906
- Gene Name
- heterogeneous nuclear ribonucleoprotein L like
- Gene Aliases
- HNRPLL, SRRF
- Chromosome Location
- Chr. 2: 38563186 - 38603036 on Build GRCh38
- UniGene ID
- Hs.445497
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142650.1 |
NP_001136122.1
|
5, 6 |
791 |
|
NM_138394.3 |
NP_612403.2
|
4, 5 |
1047 |
|
XM_005264639.3 |
XP_005264696.2
|
4, 5 |
1044 |
|
XM_005264640.3 |
XP_005264697.2
|
5, 6 |
790 |
|
XM_011533165.2 |
XP_011531467.1
|
4, 5 |
1044 |
|
XM_011533166.2 |
XP_011531468.1
|
4, 5 |
1044 |
|
GenBank mRNA |
AK000155.1 |
- |
5, 6 |
695 |
|
AK291462.1 |
- |
4, 5 |
912 |
|
AK292259.1 |
- |
5, 6 |
784 |
|
AL512692.1 |
- |
3, 4 |
441 |
|
AY236963.2 |
- |
5, 6 |
636 |
|
BC008217.1 |
- |
5, 6 |
684 |
3449607 |
BC017480.1 |
- |
4, 5 |
893 |
3502860 |
KU178839.1 |
- |
4, 5 |
637 |
|
View Full Product Details