Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr. 12: 1791957 - 1922606 on Build GRCh38
UniGene ID
Hs.13768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172364.4 NP_758952.4 11, 12 1499
XM_011521041.2 XP_011519343.1 11, 12 5410
GenBank mRNA AF516695.1 - 11, 12 1631
BC150186.1 - 11, 12 1432 40148321
BX537436.1 - 11, 12 1462
BX537437.1 - 11, 12 1500

View Full Product Details