Gene Transcripts
- Gene Symbol
- OSR2
- Entrez Gene ID
- 116039
- Gene Name
- odd-skipped related transciption factor 2
- Gene Aliases
- Chromosome Location
- Chr. 8: 98944403 - 98952104 on Build GRCh38
- UniGene ID
- Hs.253247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142462.2 |
NP_001135934.1
|
2, 3 |
1182 |
|
NM_001286841.1 |
NP_001273770.1
|
3, 4 |
1431 |
|
NM_053001.3 |
NP_443727.2
|
2, 3 |
1182 |
|
XM_005250778.4 |
XP_005250835.1
|
3, 4 |
1390 |
|
XM_011516825.2 |
XP_011515127.1
|
3, 4 |
1197 |
|
XM_011516826.2 |
XP_011515128.1
|
2, 3 |
823 |
|
XM_011516827.2 |
XP_011515129.1
|
2, 3 |
930 |
|
XM_017013018.1 |
XP_016868507.1
|
2, 3 |
802 |
|
GenBank mRNA |
AK074518.1 |
- |
2, 3 |
1143 |
|
AK291491.1 |
- |
2, 3 |
1143 |
|
AK304654.1 |
- |
3, 4 |
1175 |
|
AK310994.1 |
- |
2, 3 |
1143 |
|
AY038072.1 |
- |
2, 3 |
962 |
|
AY038073.1 |
- |
2, 3 |
1144 |
|
BC016936.1 |
- |
2, 3 |
1128 |
4425397 |
View Full Product Details