Gene Transcripts

Gene Symbol
DIRC1
Entrez Gene ID
116093
Gene Name
disrupted in renal carcinoma 1
Gene Aliases
Chromosome Location
Chr. 2: 188733738 - 188790104 on Build GRCh38
UniGene ID
Hs.470892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052952.2 NP_443184.1 2 528
XM_017003281.1 XP_016858770.1 2 349
GenBank mRNA AY039011.1 - 2 529
BC125137.1 - 1 285 40119557

View Full Product Details